This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04486380_m1
See other IL6 GE Assays ›
Gene Symbol:
Il6
Gene Name:
interleukin 6
Gene Aliases:
I79_015109
Chromosome Location:
Chr.Un|NW_003614383: 28740 - 33111 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04486380_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003507441.2 XP_003507489.1 3 - 4 425 69
GenBank mRNA AF044667.1 - 1 - 2 45 69
JI874827.1 - 3 - 4 433 69
JP057787.1 - 3 - 4 433 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Apoptosis Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0