This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04482546_m1
See other NDUFA7 GE Assays ›
Gene Symbol:
Ndufa7
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003614895: 393760 - 405337 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04482546_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003510506.2 XP_003510554.1 3 - 4 319 68
GenBank mRNA JI884439.1 - 1 - 2 156 68
JP046043.1 - 1 - 2 156 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Transporter 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0