This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04479207_m1
See other ZNF143 GE Assays ›
Gene Symbol:
Znf143
Gene Name:
zinc finger protein 143
Gene Aliases:
I79_019655, Zfp143
Chromosome Location:
Chr.Un|NW_003615051: 51816 - 83338 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04479207_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Znf143
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003511181.2 XP_003511229.1 14 - 15 1785 58
XM_007651724.1 XP_007649914.1 14 - 15 1770 58
XM_007651725.1 XP_007649915.1 14 - 15 1767 58
XM_007651726.1 XP_007649916.1 14 - 15 1780 58
GenBank mRNA JI889373.1 - 4 - 5 645 58
JP062709.1 - 4 - 5 644 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0