This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04466526_m1
See other ABRACL GE Assays ›
Gene Symbol:
Abracl
Gene Name:
ABRA C-terminal like
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613627: 656642 - 666740 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04466526_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abracl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003496538.2 XP_003496586.1 2 - 3 347 103
XM_007651099.1 XP_007649289.1 2 - 3 94 103
XM_007651104.1 XP_007649294.1 2 - 3 163 103
GenBank mRNA JI866686.1 - 2 - 3 268 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_003496538.2, XM_007651104.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0