This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04464353_m1
See other ACVR1C GE Assays ›
Gene Symbol:
Acvr1c
Gene Name:
activin A receptor, type IC
Gene Aliases:
I79_010194
Chromosome Location:
Chr.Un|NW_003613966: 1028618 - 1071030 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04464353_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acvr1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007644007.1 XP_007642197.1 6 - 7 1150 78
XM_007644008.1 XP_007642198.1 5 - 6 910 78
XM_007644009.1 XP_007642199.1 4 - 5 679 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Protein Kinase Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0