Assay Details
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007631243.1 | XP_007629433.1 | 7 - 8 | 1287 | 61 | |
GenBank mRNA | JI876868.1 | - | 2 - 3 | 331 | 61 | |
JP037277.1 | - | 2 - 3 | 331 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
