This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04441942_m1
See other MRPL19 GE Assays ›
Gene Symbol:
Mrpl19
Gene Name:
mitochondrial ribosomal protein L19
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613675: 618822 - 625848 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04441942_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003498000.2 XP_003498048.1 1 - 2 279 66
XM_007620885.1 XP_007619075.1 1 - 2 279 66
GenBank mRNA JI863624.1 - 1 - 2 237 66
JP041844.1 - 1 - 2 237 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Endogenous Controls Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0