Assay Details
Gene Symbol: Sepp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256878.1 | NP_001243807.1 | 4 - 5 | 716 | 88 | |
GenBank mRNA | JI884822.1 | - | 4 - 5 | 716 | 88 | |
JP052462.1 | - | 4 - 5 | 716 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
