This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg01354010_m1
See other NR4A3 GE Assays ›
Gene Symbol:
Nr4a3
Gene Name:
nuclear receptor subfamily 4, group A, member 3
Gene Aliases:
I79_013437
Chromosome Location:
Chr.Un|NW_003614215: 437608 - 471652 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg01354010_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nr4a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007646621.1 XP_007644811.1 5 - 6 1274 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Nuclear Receptor Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0