Assay Details
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504901.2 | XP_003504949.1 | 1 - 2 | 312 | 90 | |
GenBank mRNA | JI866392.1 | - | 1 - 2 | 279 | 90 | |
JP043330.1 | - | 1 - 2 | 279 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
