This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04947482_m1
See other PDCD1 GE Assays ›
Gene Symbol:
PDCD1
Gene Name:
programmed cell death 1
Gene Aliases:
PD-1
Chromosome Location:
Chr.25: 51606559 - 51618903 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04947482_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDCD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001314097.1 NP_001301026.1 1 - 2 117 66
XM_022409577.1 XP_022265285.1 1 - 2 305 66
GenBank mRNA AB850882.1 - 1 - 2 117 66
AB898677.1 - 1 - 2 126 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0