This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04947220_g1
See other H2AZ1 GE Assays ›
Gene Symbol:
H2AZ1
Gene Name:
H2A.Z variant histone 1
Gene Aliases:
H2AFZ
Chromosome Location:
Chr.32: 21889890 - 21892165 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04947220_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H2AZ1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287092.1 NP_001274021.1 4 - 5 502 83
XM_022413389.1 XP_022269097.1 3 - 4 386 83
GenBank mRNA CO594853.1 - 1 - 2 437 83
DN393040.1 - 4 - 5 84
GR900857.1 - 4 - 5 422 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

histone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0