This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04947191_m1
See other CEACAM16 GE Assays ›
Gene Symbol:
CEACAM16
Gene Name:
CEA cell adhesion molecule 16, tectorial membrane component
Gene Aliases:
-
Chromosome Location:
Chr.1: 110652362 - 110661173 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04947191_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286998.1 NP_001273927.1 3 - 4 660 75
XM_022422154.1 XP_022277862.1 3 - 4 1499 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0