This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04947167_m1
See other LGALS12 GE Assays ›
Gene Symbol:
LGALS12
Gene Name:
galectin 12
Gene Aliases:
-
Chromosome Location:
Chr.18: 53428894 - 53446767 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04947167_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LGALS12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284478.1 NP_001271407.1 2 - 3 160 55
XM_005631583.3 XP_005631640.2 2 - 3 572 55
XM_005631584.3 XP_005631641.2 2 - 3 574 55
XM_005631585.3 XP_005631642.1 2 - 3 574 55
XM_014120875.2 XP_013976350.2 2 - 3 241 55
XM_022405080.1 XP_022260788.1 2 - 3 573 55
XM_022405081.1 XP_022260789.1 2 - 3 574 55
XM_022405082.1 XP_022260790.1 2 - 3 573 55
XM_540894.5 XP_540894.2 2 - 3 570 55
GenBank mRNA HQ637391.1 - 2 - 3 160 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

extracellular matrix protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0