This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04649305_s1
See other MIR8902 GE Assays ›
Gene Symbol:
MIR8902 FABP6
Gene Name:
microRNA mir-8902
fatty acid binding protein 6
Gene Aliases:
cfa-mir-8902
-
Chromosome Location:
Chr.4: 50474008 - 50474112 on Build CanFam3.1
Chromosome Location:
Chr.4: 50471988 - 50477915 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf04649305_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR8902
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_128873.1 1 57 86
Gene Symbol: FABP6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022417741.1 XP_022273449.1 3 1022 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SH3 and PX domains 2B SH3PXD2B -30.1kb Sense Chr. 4 - 39854213 - 39950704 on NCBI Build CanFam3.1
Downstream ubiquitin domain containing 2 UBTD2 11.6kb Sense Chr. 4 - 39992560 - 40064512 on NCBI Build CanFam3.1

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header