This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04648793_s1
See other MIR8860 GE Assays ›
Gene Symbol:
MIR8860
Gene Name:
microRNA mir-8860
Gene Aliases:
cfa-mir-8860
Chromosome Location:
Chr.18: 47168318 - 47168452 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04648793_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR8860
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_128811.1 1 90 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream calcium voltage-gated channel auxiliary subunit alpha2delta 1 CACNA2D1 Sense Chr. 18 - 21561091 - 21949970 on NCBI Build CanFam3.1
Downstream protein piccolo-like LOC111090913 481.0kb Sense Chr. 18 - 22317374 - 22382197 on NCBI Build CanFam3.1

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0