This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04648339_s1
See other MIR8800 GE Assays ›
Gene Symbol:
MIR8800
Gene Name:
microRNA mir-8800
Gene Aliases:
cfa-mir-8800
Chromosome Location:
Chr.20: 37964390 - 37964514 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04648339_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR8800
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_128740.1 1 52 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream basic helix-loop-helix family member e40 BHLHE40 BHLHB2,
DEC1
-130.3kb Sense Chr. 20 - 12641872 - 12647519 on NCBI Build CanFam3.1
Downstream sulfatase modifying factor 1 SUMF1 315.4kb Anti-sense Chr. 20 - 13093427 - 13190621 on NCBI Build CanFam3.1

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0