Assay Details
Gene Symbol: MIR8810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128750.1 | 1 | 52 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | adrenomedullin | ADM | -170.9kb | Sense | Chr. 21 - 33472197 - 33474472 on NCBI Build CanFam3.1 | |
Downstream | ring finger protein 141 | RNF141 | 8.1kb | Anti-sense | Chr. 21 - 33653532 - 33695142 on NCBI Build CanFam3.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
