This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04647755_s1
See other MIR578 GE Assays ›
Gene Symbol:
MIR578
Gene Name:
microRNA mir-578
Gene Aliases:
cfa-mir-578
Chromosome Location:
Chr.1: 122413078 - 122413146 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04647755_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR578
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_049503.1 1 39 67

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream QKI,
KH domain containing RNA binding
QKI CQKI,
QKI-5
-901.4kb Sense Chr. 1 - 51800294 - 51956891 on NCBI Build CanFam3.1
Downstream chromosome 1 C6orf118 homolog C1H6orf118 558.2kb Anti-sense Chr. 1 - 53416602 - 53445989 on NCBI Build CanFam3.1

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header