This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04419463_gH
See other GAPDH GE Assays ›
Gene Symbol:
GAPDH
Gene Name:
glyceraldehyde-3-phosphate dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.27: 38468889 - 38472653 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04419463_gH
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GAPDH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003142.2 NP_001003142.2 5 - 6 382 54
GenBank mRNA AB038240.1 - 1 320 54
AF091136.1 - 1 98 54
DN438402.1 - 5 - 6 382 54
DQ403060.1 - 3 - 4 245 54

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
glucose metabolic process
glycolytic process
regulation of translation
peptidyl-cysteine S-trans-nitrosylation
protein stabilization
neuron apoptotic process
oxidation-reduction process
glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
microtubule binding
peptidyl-cysteine S-nitrosylase activity
NADP binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0