This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04245254_m1
See other CENPS GE Assays ›
Gene Symbol:
CENPS
Gene Name:
centromere protein S
Gene Aliases:
APITD1
Chromosome Location:
Chr.2: 85379378 - 85389175 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04245254_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CENPS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204929.1 NP_001191858.1 3 - 4 207 91
XM_005617935.3 XP_005617992.1 3 - 4 313 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

neuropeptide

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0