Assay Details
Gene Symbol: GDF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168013.1 | NP_001161485.1 | 1 - 2 | 381 | 97 | |
XM_005626358.3 | XP_005626415.1 | 2 - 3 | 830 | 97 | ||
XM_022424888.1 | XP_022280596.1 | 2 - 3 | 633 | 97 | ||
XM_022424889.1 | XP_022280597.1 | 2 - 3 | 497 | 97 | ||
XM_022424890.1 | XP_022280598.1 | 2 - 3 | 434 | 97 | ||
GenBank mRNA | GQ983543.1 | - | 1 - 2 | 381 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
