This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf03054204_m1
See other CEACAM28 GE Assays ›
Gene Symbol:
CEACAM28
Gene Name:
carcinoembryonic antigen-related cell adhesion molecule 28
Gene Aliases:
CEACAM21, CEACAM30
Chromosome Location:
Chr.1: 111812571 - 111834985 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf03054204_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM28
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097546.1 NP_001091015.1 3 - 4 736 160
XM_022422209.1 XP_022277917.1 4 - 5 1020 160
GenBank mRNA EF137910.1 - 3 - 4 736 160

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0