This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf03042918_m1
See other MIP GE Assays ›
Gene Symbol:
MIP
Gene Name:
major intrinsic protein of lens fiber
Gene Aliases:
-
Chromosome Location:
Chr.10: 701385 - 707005 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf03042918_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001313795.1 NP_001300724.1 392 57
NR_132651.1 392 57
XM_022424010.1 XP_022279718.1 1246 57
GenBank mRNA DN863767.1 - 392 57
DN864176.1 - 140 57
EF191197.1 - 392 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

lens development in camera-type eye
water transport
visual perception
positive regulation of cell adhesion
response to stimulus
protein homotetramerization
transmembrane transport
gap junction-mediated intercellular transport
structural constituent of eye lens
calmodulin binding
water channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header