This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf03034163_s1
See other CCR5 GE Assays ›
Gene Symbol:
CCR5
Gene Name:
C-C motif chemokine receptor 5
Gene Aliases:
GPCR
Chromosome Location:
Chr.20: 42293288 - 42297952 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf03034163_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCR5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012342.3 NP_001012342.2 2 1026 157
XM_022406244.1 XP_022261952.1 2 1124 157
GenBank mRNA AB183192.1 - 1 974 157
AY904725.1 - 2 1027 157

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
G-protein coupled receptor signaling pathway
cell-cell signaling
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
chemokine-mediated signaling pathway
response to cholesterol
cellular response to lipopolysaccharide
negative regulation of macrophage apoptotic process
actin binding
C-C chemokine receptor activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0