This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02734012_u1
See other H6PD GE Assays ›
Gene Symbol:
H6PD
Gene Name:
hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.5: 62548136 - 62577346 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02734012_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H6PD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005620492.2 XP_005620549.1 933 122
XM_005620495.3 XP_005620552.1 1617 122
XM_005620497.2 XP_005620554.1 970 122
XM_014113912.2 XP_013969387.1 1517 122
XM_022419026.1 XP_022274734.1 1547 122
XM_022419027.1 XP_022274735.1 970 122
XM_546762.6 XP_546762.3 853 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0