This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02730232_m1
See other MYH7 GE Assays ›
Gene Symbol:
MYH7
Gene Name:
myosin heavy chain 7
Gene Aliases:
-
Chromosome Location:
Chr.8: 3660155 - 3677650 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02730232_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYH7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113711.1 NP_001107183.1 1 - 2 897 136

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
regulation of heart rate
skeletal muscle contraction
adult heart development
regulation of the force of skeletal muscle contraction
transition between fast and slow fiber
cardiac muscle hypertrophy in response to stress
muscle filament sliding
regulation of slow-twitch skeletal muscle fiber contraction
ATP metabolic process
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
microfilament motor activity
calmodulin binding
ATP binding
actin-dependent ATPase activity
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0