This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02727665_g1
See other TAF8 GE Assays ›
Gene Symbol:
TAF8
Gene Name:
TATA-box binding protein associated factor 8
Gene Aliases:
TBN
Chromosome Location:
Chr.12: 10689268 - 10720945 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02727665_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAF8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005627343.3 XP_005627400.1 3 - 4 328 120
XM_005627344.3 XP_005627401.1 3 - 4 328 120
XM_005627346.3 XP_005627403.1 3 - 4 328 120
XM_022426041.1 XP_022281749.1 3 - 4 328 120
XM_022426042.1 XP_022281750.1 3 - 4 458 120
XM_538919.6 XP_538919.3 3 - 4 328 120

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0