This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02726818_m1
See other CFAP91 GE Assays ›
Gene Symbol:
CFAP91
Gene Name:
cilia and flagella associated protein 91
Gene Aliases:
C33H3orf15, MAATS1
Chromosome Location:
Chr.33: 23315184 - 23447307 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02726818_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CFAP91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005639531.3 XP_005639588.1 12 - 13 1631 92
XM_005639532.3 XP_005639589.1 12 - 13 1632 92
XM_014110238.2 XP_013965713.1 11 - 12 1489 92
XM_014110239.2 XP_013965714.1 13 - 14 1651 92
XM_022413976.1 XP_022269684.1 13 - 14 1617 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header