Assay Details
Gene Symbol: GAREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421721.1 | XP_022277429.1 | 5 - 6 | 580 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: GAREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421721.1 | XP_022277429.1 | 5 - 6 | 580 | 146 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |