This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02720142_u1
See other MOBP GE Assays ›
Gene Symbol:
MOBP
Gene Name:
myelin associated oligodendrocyte basic protein
Gene Aliases:
-
Chromosome Location:
Chr.23: 9121277 - 9176128 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02720142_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005634267.2 XP_005634324.1 4 529 140
XM_014106596.2 XP_013962071.1 4 529 140
XM_014106597.2 XP_013962072.1 4 529 140
XM_014106598.2 XP_013962073.1 5 596 140
XM_014106599.2 XP_013962074.1 4 529 140
XM_014106600.2 XP_013962075.1 3 432 140
XM_014106601.2 XP_013962076.1 4 529 140
XM_022408499.1 XP_022264207.1 4 529 140

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

myelin protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0