This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02719629_m1
See other MRPL52 GE Assays ›
Gene Symbol:
MRPL52
Gene Name:
mitochondrial ribosomal protein L52
Gene Aliases:
-
Chromosome Location:
Chr.8: 3177010 - 3181526 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02719629_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPL52
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005623210.2 XP_005623267.1 4 - 5 327 164
XM_005623212.2 XP_005623269.1 4 - 5 333 164
XM_005623214.2 XP_005623271.1 4 - 5 291 164
XM_005623215.3 XP_005623272.1 2 - 3 198 164
XM_022421798.1 XP_022277506.1 3 - 4 171 164
XM_537363.5 XP_537363.3 4 - 5 286 164

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0