This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02716842_s1
See other MYL1 GE Assays ›
Gene Symbol:
MYL1
Gene Name:
myosin light chain 1
Gene Aliases:
-
Chromosome Location:
Chr.37: 18143644 - 18168208 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02716842_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003434275.4 XP_003434323.1 4 357 150
XM_005640612.3 XP_005640669.1 5 453 150
XM_005640613.3 XP_005640670.1 4 560 150
XM_005640614.2 XP_005640671.1 4 357 150
XM_014110930.2 XP_013966405.1 5 453 150
XM_022415004.1 XP_022270712.1 4 345 150
XM_022415005.1 XP_022270713.1 4 345 150
XM_536054.6 XP_536054.2 4 560 150

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0