This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02714493_m1
See other PRR5L GE Assays ›
Gene Symbol:
PRR5L
Gene Name:
proline rich 5 like
Gene Aliases:
-
Chromosome Location:
Chr.18: 31708888 - 31849365 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02714493_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRR5L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005631111.3 XP_005631168.1 7 - 8 1200 137
XM_005631112.3 XP_005631169.1 6 - 7 1050 137
XM_014121216.2 XP_013976691.1 7 - 8 1497 137
XM_022405314.1 XP_022261022.1 8 - 9 1588 137
XM_022405315.1 XP_022261023.1 11 - 12 2272 137
XM_022405316.1 XP_022261024.1 11 - 12 2186 137
XM_022405317.1 XP_022261025.1 6 - 7 699 137
XM_022405319.1 XP_022261027.1 3 - 4 961 137
XM_022405320.1 XP_022261028.1 4 - 5 1012 137
XM_845585.4 XP_850678.1 6 - 7 800 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0