This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02704244_s1
See other GJA8 GE Assays ›
Gene Symbol:
GJA8
Gene Name:
gap junction protein alpha 8
Gene Aliases:
-
Chromosome Location:
Chr.17: 58360313 - 58428751 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02704244_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJA8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005630710.3 XP_005630767.1 7 947 165
XM_005630711.3 XP_005630768.1 6 806 165
XM_022404918.1 XP_022260626.1 7 1009 165
XM_022404919.1 XP_022260627.1 7 956 165
XM_022404920.1 XP_022260628.1 6 915 165
XM_022404921.1 XP_022260629.1 8 1177 165
XM_022404922.1 XP_022260630.1 6 870 165
XM_022404923.1 XP_022260631.1 6 743 165
XM_022404924.1 XP_022260632.1 5 722 165
XM_022404925.1 XP_022260633.1 5 628 165
XM_022404926.1 XP_022260634.1 4 487 165
XM_022404927.1 XP_022260635.1 7 946 165
XM_022404928.1 XP_022260636.1 7 946 165

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

gap junction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0