This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02703088_s1
See other ESRRA GE Assays ›
Gene Symbol:
ESRRA
Gene Name:
estrogen related receptor alpha
Gene Aliases:
ESTRRA
Chromosome Location:
Chr.18: 52755547 - 52762938 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02703088_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESRRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002936.1 NP_001002936.1 1 74 150
XM_005631023.2 XP_005631080.1 2 230 150
XM_022405056.1 XP_022260764.1 2 478 150
GenBank mRNA AY533243.1 - 1 74 150

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
steroid hormone mediated signaling pathway
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
steroid binding
zinc ion binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0