Assay Details
Gene Symbol: STAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634223.3 | XP_005634280.1 | 10 - 11 | 1299 | 135 | |
XM_005634224.3 | XP_005634281.1 | 9 - 10 | 1107 | 135 | ||
XM_022408442.1 | XP_022264150.1 | 10 - 11 | 1303 | 135 | ||
XM_534217.5 | XP_534217.2 | 9 - 10 | 1140 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
