Assay Details
Gene Symbol: RPH3AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423441.1 | XP_022279149.1 | 5 - 6 | 499 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: RPH3AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423441.1 | XP_022279149.1 | 5 - 6 | 499 | 100 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |