This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02699110_m1
See other PAICS GE Assays ›
Gene Symbol:
PAICS
Gene Name:
phosphoribosylaminoimidazole carboxylase and phosphoribosylaminoimidazolesuccinocarboxamide synthase
Gene Aliases:
KIAA1211
Chromosome Location:
Chr.13: 48296232 - 48682650 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02699110_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PAICS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003639458.4 XP_003639506.1 8 - 9 1110 77
XM_022427005.1 XP_022282713.1 21 - 22 6011 77
XM_022427006.1 XP_022282714.1 20 - 21 5903 77
XM_022427008.1 XP_022282716.1 24 - 25 6404 77
XM_022427009.1 XP_022282717.1 21 - 22 5913 77
XM_022427010.1 XP_022282718.1 20 - 21 5696 77
XM_022427011.1 XP_022282719.1 23 - 24 6268 77
XM_022427012.1 XP_022282720.1 19 - 20 5630 77
XM_022427013.1 XP_022282721.1 22 - 23 6103 77
XM_022427015.1 XP_022282723.1 14 - 15 2418 77
XM_022427016.1 XP_022282724.1 14 - 15 2320 77

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header