This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02695336_m1
See other BAK1 GE Assays ›
Gene Symbol:
BAK1
Gene Name:
BCL2 antagonist/killer 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 2950359 - 2957604 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02695336_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BAK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001020808.1 NP_001018644.1 4 - 5 543 114
XM_005627072.3 XP_005627129.1 5 - 6 810 114
XM_022425734.1 XP_022281442.1 5 - 6 822 114
XM_022425735.1 XP_022281443.1 5 - 6 674 114
XM_022425736.1 XP_022281444.1 5 - 6 587 114
XM_022425737.1 XP_022281445.1 4 - 5 1167 114
XM_022425738.1 XP_022281446.1 4 - 5 498 114
GenBank mRNA DQ002813.1 - 4 - 5 543 114

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0