This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02695123_m1
See other SLC2A4 GE Assays ›
Gene Symbol:
SLC2A4
Gene Name:
solute carrier family 2 member 4
Gene Aliases:
GLUT4
Chromosome Location:
Chr.5: 32238095 - 32244022 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02695123_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC2A4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159327.1 NP_001152799.1 8 - 9 1208 91
XM_005619478.3 XP_005619535.3 8 - 9 1096 91
GenBank mRNA AJ388533.1 - 1 - 2 85 91
CX008986.1 - 1 - 2 138 91

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cellular response to insulin stimulus
glucose import in response to insulin stimulus
glucose transmembrane transport
glucose transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0