This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02694373_m1
See other DDX19B GE Assays ›
Gene Symbol:
DDX19B
Gene Name:
DEAD-box helicase 19B
Gene Aliases:
-
Chromosome Location:
Chr.5: 76314675 - 76336318 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02694373_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DDX19B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005620703.3 XP_005620760.1 2 - 3 160 153
XM_856865.5 XP_861958.3 2 - 3 247 153

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0