Assay Details
Gene Symbol: ZNF280D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638445.3 | XP_005638502.1 | 15 - 16 | 2028 | 112 | |
XM_005638446.2 | XP_005638503.1 | 13 - 14 | 1960 | 112 | ||
XM_005638448.3 | XP_005638505.1 | 15 - 16 | 2036 | 112 | ||
XM_014109457.2 | XP_013964932.1 | 15 - 16 | 2036 | 112 | ||
XM_014109458.2 | XP_013964933.1 | 16 - 17 | 2837 | 112 | ||
XM_014109460.2 | XP_013964935.1 | 10 - 11 | 2921 | 112 | ||
XM_014109462.2 | XP_013964937.1 | 15 - 16 | 2036 | 112 | ||
XM_022412620.1 | XP_022268328.1 | 16 - 17 | 2844 | 112 | ||
XM_022412621.1 | XP_022268329.1 | 15 - 16 | 2775 | 112 | ||
XM_022412622.1 | XP_022268330.1 | 14 - 15 | 1970 | 112 | ||
XM_022412623.1 | XP_022268331.1 | 11 - 12 | 1755 | 112 | ||
XM_022412624.1 | XP_022268332.1 | 11 - 12 | 1707 | 112 | ||
XM_022412625.1 | XP_022268333.1 | 15 - 16 | 2036 | 112 | ||
XM_022412626.1 | XP_022268334.1 | 15 - 16 | 2036 | 112 | ||
XM_535490.5 | XP_535490.3 | 14 - 15 | 2027 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
