This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02693103_m1
See other ADORA2A GE Assays ›
Gene Symbol:
ADORA2A
Gene Name:
adenosine A2a receptor
Gene Aliases:
-
Chromosome Location:
Chr.26: 28141076 - 28150600 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02693103_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADORA2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003278.1 NP_001003278.1 1 - 2 627 105
GenBank mRNA X14052.1 - 627 105

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

synaptic transmission, dopaminergic
adenosine receptor signaling pathway
response to amphetamine
adenylate cyclase-activating G-protein coupled receptor signaling pathway
locomotory behavior
positive regulation of synaptic transmission, GABAergic
eating behavior
negative regulation of alpha-beta T cell activation
negative regulation of inflammatory response
inhibitory postsynaptic potential
positive regulation of apoptotic signaling pathway
G-protein coupled adenosine receptor activity
enzyme binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0