This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02693010_m1
See other SNCA GE Assays ›
Gene Symbol:
SNCA
Gene Name:
synuclein alpha
Gene Aliases:
-
Chromosome Location:
Chr.32: 12915871 - 13079338 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02693010_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNCA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014110040.2 XP_013965515.1 2 - 3 322 97
XM_022413536.1 XP_022269244.1 2 - 3 329 97
XM_022413537.1 XP_022269245.1 2 - 3 329 97
XM_022413538.1 XP_022269246.1 2 - 3 317 97
XM_535656.6 XP_535656.1 2 - 3 313 97
XM_855821.5 XP_860914.1 2 - 3 303 97

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0