Assay Details
Gene Symbol: NIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618207.3 | XP_005618264.1 | 824 | 162 | ||
XM_014112285.2 | XP_013967760.1 | 966 | 162 | |||
XM_022416803.1 | XP_022272511.1 | 663 | 162 | |||
XM_536157.6 | XP_536157.2 | 767 | 162 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
