This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02691372_s1
See other CHRM3 GE Assays ›
Gene Symbol:
CHRM3
Gene Name:
cholinergic receptor muscarinic 3
Gene Aliases:
-
Chromosome Location:
Chr.4: 504663 - 999550 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02691372_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHRM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005618760.2 XP_005618817.1 1862 117
XM_005618761.2 XP_005618818.1 1866 117
XM_005618763.2 XP_005618820.1 1799 117
XM_005618766.2 XP_005618823.1 1965 117
XM_005618768.2 XP_005618825.1 1891 117
XM_005618769.2 XP_005618826.1 2029 117
XM_014112712.2 XP_013968187.1 1803 117
XM_014112713.2 XP_013968188.1 2154 117
XM_014112714.2 XP_013968189.1 1690 117
XM_022417355.1 XP_022273063.1 2072 117
XM_022417356.1 XP_022273064.1 2132 117
XM_022417357.1 XP_022273065.1 2161 117
XM_022417358.1 XP_022273066.1 2227 117
XM_022417359.1 XP_022273067.1 2118 117
XM_022417360.1 XP_022273068.1 1955 117
XM_022417361.1 XP_022273069.1 1753 117
XM_022417362.1 XP_022273070.1 1900 117
XM_022417363.1 XP_022273071.1 1883 117
XM_022417364.1 XP_022273072.1 1497 117
XM_546069.6 XP_546069.4 1916 117
GenBank mRNA AF056305.1 - 317 117

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0