This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02689177_u1
See other PROC GE Assays ›
Gene Symbol:
PROC
Gene Name:
protein C, inactivator of coagulation factors Va and VIIIa
Gene Aliases:
-
Chromosome Location:
Chr.19: 23173626 - 23183634 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02689177_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PROC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013849.1 NP_001013871.1 6 512 179
XM_005631762.3 XP_005631819.1 6 578 179
XM_014121368.2 XP_013976843.1 6 639 179
XM_014121369.2 XP_013976844.1 6 755 179
XM_022405862.1 XP_022261570.1 6 686 179
XM_022405864.1 XP_022261572.1 6 660 179

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
blood coagulation
negative regulation of apoptotic process
negative regulation of inflammatory response
negative regulation of coagulation
positive regulation of establishment of endothelial barrier
serine-type endopeptidase activity
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0