This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02686663_m1
See other VPS35L GE Assays ›
Gene Symbol:
VPS35L
Gene Name:
VPS35 endosomal protein sorting factor like
Gene Aliases:
C6H16orf62
Chromosome Location:
Chr.6: 25594735 - 25710159 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02686663_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VPS35L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005622072.3 XP_005622129.3 19 - 20 2056 95
XM_014114590.2 XP_013970065.2 19 - 20 2056 95
XM_014114591.2 XP_013970066.1 18 - 19 1652 95
XM_022420029.1 XP_022275737.1 18 - 19 1996 95
XM_022420030.1 XP_022275738.1 19 - 20 2056 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0