Assay Details
Gene Symbol: ZNF618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424969.1 | XP_022280677.1 | 15 | 1986 | 154 | |
XM_022424970.1 | XP_022280678.1 | 15 | 1983 | 154 | ||
XM_022424971.1 | XP_022280679.1 | 14 | 1950 | 154 | ||
XM_022424972.1 | XP_022280680.1 | 14 | 1941 | 109 | ||
XM_022424973.1 | XP_022280681.1 | 14 | 1938 | 109 | ||
XM_022424974.1 | XP_022280682.1 | 14 | 1926 | 154 | ||
XM_022424975.1 | XP_022280683.1 | 14 | 1890 | 154 | ||
XM_022424976.1 | XP_022280684.1 | 13 | 1890 | 154 | ||
XM_022424977.1 | XP_022280685.1 | 13 | 1881 | 109 | ||
XM_022424978.1 | XP_022280686.1 | 12 | 1785 | 109 | ||
XM_022424979.1 | XP_022280687.1 | 12 | 1782 | 109 | ||
XM_022424980.1 | XP_022280688.1 | 15 | 1557 | 154 | ||
XM_022424981.1 | XP_022280689.1 | 14 | 1746 | 154 | ||
XM_022424982.1 | XP_022280690.1 | 14 | 1743 | 154 | ||
XM_022424983.1 | XP_022280691.1 | 13 | 1701 | 109 | ||
XM_022424984.1 | XP_022280692.1 | 13 | 1698 | 109 | ||
XM_022424985.1 | XP_022280693.1 | 13 | 1683 | 154 | ||
XM_022424986.1 | XP_022280694.1 | 12 | 1602 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
